Learn More
Thermo Scientific™ AerosolSense™ Cartridges
Product Code. 17266770 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
AerosolSense Sampler
Unit Size:
Pack of 30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17266770 AerosolSense Sampler Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17266770 Supplier Thermo Scientific™ Supplier No. 12148001
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
Used with the Thermo Scientific™ AerosolSense™ Sampler
  • Small and easy to use
  • Isolated collection substrate
  • Can only be used with the Thermo Scientific™ AerosolSense™ Sampler
TRUSTED_SUSTAINABILITY
Specifications
Product Type AerosolSense™ Cartridges
For Use With (Equipment) AerosolSense Sampler
Quantity 30 Pk.