Learn More
Gardner Denver Welch™ Advanced Vacuum Oven Kit for DN 16/DN 25 Rotary Vane Pumps
Shop All Welch Ilmvac ProductsDescription
Advanced vacuum oven kit includes:
- 2.5m red rubber vacuum hose, 8mm ID, 5mm wall thickness
- KF16 flange adapter
- KF25 flange adapter
- Hose nozzle for G1/4 thread
- In-line particle filter
- Replacement filter element
- Clamps and centring rings
Specifications
Specifications
| For Use With | DN 16/DN 25 Rotary Vane Pumps |
| Product Line | Advanced Vacuum Oven Kit |