Learn More
Advanced Instruments Osmolality Linearity Set
Description
- Each level of standard supplied in the set should be run and documented to verify the reportable range of patient results
- Run the linearity set until accurate results obtained or recalibration needed
- For in vitro diagnostic use with all brands of osmometers
Specifications
Specifications
| Certifications/Compliance | CLIA, CE |
| Content And Storage | 2°C to 30°C |
| Concentration | 100, 500, 900, 1500, and 2000 mOsm/kg H2O |
| For Use With (Equipment) | 3320 Micro-osmometer |
| Type | Osmolality Linearity set |
| Intended Use | Monitoring performance of any osmometer in the range 0 to 2000mOsm/kg |
| Quantity | 10 x 5 mL |