Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Product Code. 12737839
Click to view available options
Depth (Metric) Interior:
300 mm
325 mm
505 mm
690 mm
Height (Metric) Interior:
150 mm
200 mm
Width (Metric) Interior:
150 mm
300 mm
Temperature Range:
-15°C to +200°C
0°C to 200°C
Unit Size:
Each
Product Code. 12737839
Brand: Grant Instruments™ TX150ST5
Only null left
Add to basket
Add to basket
Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150°C. Grant Instruments™ Optima™ TX150 Series Digital Heated Circulating Bath, Stainless Steel Tank combines the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of five stainless steel (ST) tanks with different capacities.
  • Series of digitally controlled heated circulating baths provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150ºC.
  • Cmbine the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of three economy plastic (P) tanks or five stainless steel (ST) tanks with different capacities
  • Heating circulator incorporates Intelligent Control Optimisation™ (ICO) for adaptive intelligent PID temperature control
  • Includes a powerful integral pump, making TX150 systems suitable both for immersing samples and circulating temperature controlled fluid to external devices 
TRUSTED_SUSTAINABILITY
Specifications
Temperature Range 0°C to 200°C
Includes Includes TX150 immersion heater/stirrer with ST5 Bath
Material Stainless Steel
Width (Metric) Interior 150 mm
Height (Metric) Interior 150 mm
Depth (Metric) Interior 300 mm