Learn More
ADU-S100, MedChemExpress
Description
ADU-S100 (MIW815), an activator of stimulator of interferon genes (STING), leads to potent and systemic tumor regression and immunity.
Specifications
Specifications
| Chemical Name or Material | ADU-S100 |
| CAS | 1638241-89-0 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H24N10O10P2S2 |
| Quantity | 1 mg |
| Synonym | MIW815 ML RR-S2 CDA |
| Solubility Information | H2O : 20 mg/mL (28.96 mM; Need ultrasonic) |
| SMILES | O[C@H]1[C@](O[P@](S)(OC[C@](O[C@](N2C3=NC=NC(N)=C3N=C2)([H])[C@@H]4O)([H])[C@@]4([H])O5)=O)([H])[C@@](N6C7=NC=NC(N)=C7N=C6)([H])O[C@]1([H])CO[P@]5(S)=O |
| Molecular Weight (g/mol) | 690.54 |
| Show More |