missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADRB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13096-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detects ADRB2 with an apparent molecular weight of 50-55 kDa as has been shown by some researches.
The beta-2 adrenoceptor (ADRB2) is an Adrenergic Receptor expressed in smooth muscle and metabolic tissues. Activation of this receptor induces a decrease in gastrointestinal motility, bronchodilation, vasodilation in skeletal and cardiac muscle, and glycogenolysis in liver. The beta-2 adrenoceptor is also a major lipolytic receptor in human fat cells and may be involved in obesity. Agonists of ADRB2 are most widely used for the treatment of asthma. In complex with beta-arrestin-1 and c-src, the beta-2 adrenergic receptor activates MAP kinases ERK1(MAPK3) and ERK2 (MAPK1). Expression of the beta-2 adrenoceptor has been reported in adipose, blood, brain, heart, lung, nose, pancreas, skeletal muscle, skin, and vessel. ESTs have been isolated from breast, liver, liver/spleen, placenta, and uterus libraries.Specifica
| ADRB2 | |
| Polyclonal | |
| Unconjugated | |
| ADRB2 | |
| ADRB2, ADRB2R, ADRBR, B2AR, BAR, Beta 2 adrenergic receptor, Beta 2 adrenoceptor, Beta 2 adrenoreceptor, BETA2AR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11555, 154, 24176 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P07550, P10608, P18762 | |
| Adrb2 | |
| ADRB2 Fusion Protein Ag3715 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto