Learn More
Melford Adonitol
Click to view available options
Quantity:
500 g
Unit Size:
500g
Description
Adonitol is a crystalline pentose alcohol formed by the reduction of ribose.
Adonitol is a crystalline pentose alcohol formed by the reduction of ribose. It occurs naturally in the plant Adonis vernalis, as well as in the cell walls of Gram positive bacteria.
Specifications
Specifications
| CAS | 488-81-3 |
| Molecular Formula | C5H12O5 |
| Quantity | 500 g |