missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ADK (Human) Recombinant Protein

Human ADK full-length ORF ( AAH03568.1, 1 a.a. - 345 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000132-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141291

  • 6270.00 NOK / 25µg
Estimated Shipment: 10-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MTSVRENILFGMGNPLLDISAVVDKDFLDKYSLKPNDQILAEDKHKELFDELVKKFKVEYHAGGSTQNSIKVAQWMIQQPHKAATFFGCIGIDKFGEILKRKAAEAHVDAHYYEQNEQPTGTCAACITGDNRSLIANLAAANCYKKEKHLDLEKNWMLVEKARVCYIAGFFLTVSPESVLKVAHHASENNRIFTLNLSAPFFSQFYKESLMKVMPYVDILFGNETEAATFAREQGFETKDIKEIAKKTQALPKMNSKRQRIVIFTQGRDDTIMATESEVTAFAVLDQDQKEIIDTNGAGDAFVGGFLSQLVSDKPLTECIRAGHYAASIIIRRTGCTFPEKPDFH
Specifications

Specifications

AAH03568.1
adenosine kinase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADK
GST
132
Wheat germ expression system
25 μg
AK
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers