Learn More
HUBER Adjustable Bypass
Click to view available options
Size:
M16 x 1
M24 x 1.5
M30 x 1.5
M38 x 1.5
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Unistat |
| Temperature Range | -20°C to +150°C |
| Type | Bypass |
| Includes | Manometer is not included. |
| Size | M30 x 1.5 |