Learn More
HUBER Adjustable Bypass
Brand:  HUBER 9970
 View more versions of this product
Product Code. 15206567
  • ONLINE EXCLUSIVE PRICE
    12330.00 NOK
    11713.87 NOK / Each
    Save 616.13 NOK
    4% Off
Specifications
Unistat
Bypass
M38 x 1.5
-20°C to +150°C
Manometer is not included.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
HUBER Adjustable Bypass > Size: M38 x 1.5