missing translation for 'onlineSavingsMsg'
Learn More

Adiponitrile, 98%, Thermo Scientific Chemicals

Catalog Number p-7026855
Click to view available options
Quantity:
250 mL
1000 mL
This item is not returnable. View return policy
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N
This item is not returnable. View return policy
111-69-3
C6H8N2
108.144
MFCD00001975
BTGRAWJCKBQKAO-UHFFFAOYSA-N
adiponitrile, 1,4-dicyanobutane, adipodinitrile, adipyldinitrile, adipic acid dinitrile, tetramethylene dicyanide, nitrile adipico, tetramethylene cyanide, adipic acid nitrile, adipinsaeuredinitril
8128
hexanedinitrile
C(CCC#N)CC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 2°C
Density 0.951
Boiling Point 295°C
Flash Point 163°C (325°F)
Odor Odorless
Refractive Index 1.438
Quantity 250 mL
UN Number UN2205
Beilstein 1740005
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material Adiponitrile
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'