Learn More
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats
Product Code. 11595315 Shop All Texwipe Products
Change view
Click to view available options
Color:
Blue
Gray
White
Quantity:
30 layer
60 layer
Unit Size:
Pack of 4
Pack of 8
Product Code. Color Quantity unitSize
11595315 Gray 30 layer Pack of 8
11555325 Blue 60 layer Pack of 4
11565325 Gray 60 layer Pack of 4
11585315 White 60 layer Pack of 4
4 options
Product Code. 11595315
Brand: ITW Chemische Produkte™ AMA184681G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Reduce contamination in high traffic areas. ITW Chemische CleanStep™ Adhesive Cleanroom Mats are constructed with either 30 or 60 layers. Each removable sheet has a consistent adhesive coating designed to trap particles and prevent transfer.
  • Designed to capture particulates from foot and wheel borne traffic prior to entry into cleanrooms or controlled environments
  • Provide a portable contamination control solution
  • Can be used with or without frames (sold separately)
  • Contain anti-microbial preservatives that prevent microbial growth
  • Consistent adhesive coating on each layer of 30 and 60 layer mats
  • Possible applications include cleanroom and critical environment entry points, gowning rooms or anterooms in the semiconductor, biotechnology, medical device, pharmaceutical processing, aerospace, and automotive markets
Specifications
Dimensions (L x W) 18 x 46 in.
Quantity 30 layer
Color Gray
Material Polyethylene