Learn More
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats
Product Code. 11555325 Shop All Texwipe Products
Change view
Click to view available options
Color:
Blue
Gray
White
Quantity:
30 layer
60 layer
Unit Size:
Pack of 4
Pack of 8
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
11555325 Blue 60 layer Pack of 4
11595315 Gray 30 layer Pack of 8
11565325 Gray 60 layer Pack of 4
11585315 White 60 layer Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11555325 Supplier ITW Chemische Produkte™ Supplier No. AMA184682B
  • ONLINE EXCLUSIVE PRICE
    2595.00 NOK
    2342.68 NOK / Pack of 4
    Save 252.32 NOK
    9% Off
View all promotions
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
Reduce contamination in high traffic areas. ITW Chemische CleanStep™ Adhesive Cleanroom Mats are constructed with either 30 or 60 layers. Each removable sheet has a consistent adhesive coating designed to trap particles and prevent transfer.
  • Designed to capture particulates from foot and wheel borne traffic prior to entry into cleanrooms or controlled environments
  • Provide a portable contamination control solution
  • Can be used with or without frames (sold separately)
  • Contain anti-microbial preservatives that prevent microbial growth
  • Consistent adhesive coating on each layer of 30 and 60 layer mats
  • Possible applications include cleanroom and critical environment entry points, gowning rooms or anterooms in the semiconductor, biotechnology, medical device, pharmaceutical processing, aerospace, and automotive markets
Specifications
Dimensions (L x W) 18 x 46 in.
Quantity 60 layer
Color Blue
Material Polyethylene