Learn More
ADH4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16474-1-AP-150UL
 View more versions of this product
Product Code. 16896104
  • 4836.10 NOK / 150µL
Description
This gene encodes class II alcohol dehydrogenase 4 pi subunit, which is a member of the alcohol dehydrogenase family. Members of this enzyme family metabolize a wide variety of substrates, including ethanol, retinol, other aliphatic alcohols, hydroxysteroids, and lipid peroxidation products. Class II alcohol dehydrogenase is a homodimer composed of 2 pi subunits. It exhibits a high activity for oxidation of long-chain aliphatic alcohols and aromatic alcohols and is less sensitive to pyrazole. This gene is localized to chromosome 4 in the cluster of alcohol dehydrogenase genes.
Specifications
Specifications
ADH4
Polyclonal
Unconjugated
ADH4
ADH 2, ADH4, Alcohol dehydrogenase 4
Rabbit
Antigen Affinity Chromatography
RUO
127, 26876, 29646
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08319, Q64563, Q9QYY9
ADH4
ADH4 Fusion Protein Ag9568
150 μL
Primary
Human, Mouse, Rat