Learn More
3M™ Adflo™ Storage Bag
Product Code. 15899634 Shop All 3M Products
Change view
Click to view available options
Closure Type:
Pull cord, zip lock
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type Unit Size
15899634 Pull cord, zip lock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15899634 Supplier 3M™ Supplier No. 7000044376
  • ONLINE EXCLUSIVE PRICE
    506.00 NOK
    309.64 NOK / Pack of 1
    Save 196.36 NOK
    38% Off
Estimated Shipment: 15-01-2027
3M™ Adflo™ Storage Bag is a replacement protective bag for Adflo™ Turbo.
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
TRUSTED_SUSTAINABILITY
Specifications
Product Type Storage Bag
Closure Type Pull cord, zip lock
Color Black
For Use With (Equipment) Adflo™ Turbo