missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine A2aR/A2bR Antibody (599743) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB94972JF669
This item is not returnable.
View return policy
Description
Adenosine A2aR/A2bR Monoclonal antibody specifically detects Adenosine A2aR/A2bR in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Adenosine A2aR/A2bR | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 599743 | |
| 50mM Sodium Borate | |
| NS0 mouse myeloma cell line transfected with human Adenosine A2aR, Met1-Ser412, Accession # P29274 | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction