Learn More
Adenosine 5'-Monophosphate, TRC
Product Code. 30135275
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135275 50 g 50g
30138739 25 g 25g
30008017 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135275 Supplier Toronto Research Chemicals Supplier No. TRCA28178050G
 Restricted Item
Estimated Shipment: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61-19-8
Molecular Formula C10 H14 N5 O7 P
Synonym 5'-Adenylic acid∣5'-AMP∣AMP∣AMP (nucleotide)∣Adenosine 5'-(dihydrogen phosphate)∣Adenosine 5'-monophosphate∣Adenosine 5'-phosphate∣Adenosine 5'-phosphoric acid∣Adenosine monophosphate∣Adenosine phosphate∣Adenosine-5-monophosphoric acid∣Adenosine-5'-monophosphoric acid∣Adenovite∣Adenylic acid∣Cardiomone∣Lycedan∣My-B-Den∣NSC 20264∣Phosaden∣Phosphaden∣Phosphentaside
Specifications
Format Neat
Molecular Weight (g/mol) 347.22
Product Type API
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 50 g
Chemical Name or Material 5'-Adenylic Acid