Learn More
Adenosine 5'-Diphosphate Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Adenosine 5'-Diphosphate Sodium Salt |
| CAS | 20398-34-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10H15N5O10P2 . x(Na) |
| Quantity | 2.5 g |
| Synonym | Adenosine 5-Diphosphate Monosodium Salt |
| InChI Formula | InChI=1S/C10H15N5O10P2/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(24-10)1-23-27(21,22)25-26(18,19)20/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H2,11,12,13)(H2,18,19,20)/t4-,6-,7-,10-/m1/s1 |
| SMILES | [Na+].Nc1ncnc2c1ncn2[C@@H]3O[C@H](COP(=O)(O)OP(=O)(O)[O-])[C@@H](O)[C@H]3O |
| IUPAC Name | [(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphono hydrogen phosphate |
| Molecular Weight (g/mol) | 427.2 |
| Show More |