missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-Diphosphate Monopotassium Salt Dihydrate, MP Biomedicals™

Product Code. 11483071 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 11483071

Brand: MP Biomedicals 02150259.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acts as a P2Y receptor agonist. Adenosine-5'-Diphosphate Monopotassium Salt Dihydrate, MP Biomedicals has been used to study the release, neuronal effects and removal of extracellular β-NAD+ and to compare prasugrel versus clopidogrel antiplatelet th

Adenosine 5′-diphosphate has been used to study the release, neuronal effects and removal of extracellular β-nicotinamide adenine dinucleotide (β-NAD+) in the rat brain. Adenosine 5′-diphosphate has also been used to compare prasugrel versus clopidogrel antiplatelet therapy after acute coronary syndrome.

TRUSTED_SUSTAINABILITY

Specifications

CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
MDL Number MFCD00066472
Quantity 500 mg
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
Solubility Information Soluble in water (5mg/mL, clear and colorless solution).
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
IUPAC Name potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
Molecular Weight (g/mol) 465.29
PubChem CID 131675483
Formula Weight 501.2
Percent Purity >98%
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.