Learn More
Thermo Scientific Chemicals Adenosine-5'-diphosphate monopotassium salt dihydrate, 99%
Product Code. 15420147
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15420147 1 g 1g
15430147 5 g 5g
2 options
Product Code. 15420147
Brand: Thermo Scientific Chemicals J60672.03
Request Bulk or Custom Format
CAS: 72696-48-1 | C10H14KN5O10P2 | 465.29 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
IUPAC Name potassium;(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolan-3-olate;dihydrate
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Specifications
Color White
Quantity 1 g
Formula Weight 501.33 (465.29 Anhydrous)
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphosphate monopotassium salt dihydrate