Learn More
Adenosine-5'-diphophate monopotassium salt, 98 to 100%, MP Biomedicals™
Product Code. 11493071 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11493071
Brand: MP Biomedicals 0215025901
Chemical Identifiers
CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1
Specifications
Color White
Assay Percent Range 98 to 100%
Quantity 1 g
IUPAC Name potassium;(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-4-hydroxy-2-[[hydroxy(phosphonooxy)phosphoryl]oxymethyl]oxolan-3-olate;dihydrate
Formula Weight 465.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphophate monopotassium salt