missing translation for 'onlineSavingsMsg'
Learn More

Adenosine-5'-diphophate monopotassium salt, 98 to 100%, MP Biomedicals™

Product Code. 11403081 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11403081 5 g 5g
11493071 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11403081

Brand: MP Biomedicals 0215025905

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 72696-48-1
Molecular Formula C10H14KN5O10P2
Molecular Weight (g/mol) 465.29
MDL Number MFCD00066472
InChI Key ZNCWUOPIJTUALR-GWKNMROSNA-M
Synonym adenosine 5-diphosphate monopotassium salt dihydrate
PubChem CID 131675483
SMILES [K+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
Assay Percent Range 98 to 100%
Quantity 5 g
IUPAC Name potassium (2R,3R,4S,5R)-2-(6-amino-9H-purin-9-yl)-5-({[(hydrogen phosphonatooxy)(hydroxy)phosphoryl]oxy}methyl)oxolane-3,4-diol
Formula Weight 465.2
Physical Form Crystalline Powder
Chemical Name or Material Adenosine-5'-diphophate monopotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.