missing translation for 'onlineSavingsMsg'
Learn More

Adenine Hydrochloride, ∼99%, MP Biomedicals™

Product Code. 11398509 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11398509

Brand: MP Biomedicals 0210019105

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adenine Hydrochloride, ∼99% is used in viral research and has a wide range of chemical and biochemical roles in vivo and in vitro.

  • Adenine has a wide range of chemical and biochemical roles in vivo and in vitro.
  • It is a regulatory molecule and a component of DNA, RNA, cofactors (NAD, FAD) and signaling molecules (cAMP).
  • It is used as a blood preservative and in viral research.
  • It is a purine nucleobase widespread throughout animal and plant tissues combined with niacinamide, D-ribose, and phosphoric acids.
  • It is a constituent of nucleic acids and coenzymes, such as codehydrase I and II, adenylic acid, coalaninedehydrase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-72-7
Molecular Formula C5H8ClN5O
Molecular Weight (g/mol) 189.60
MDL Number MFCD11046306
InChI Key MYRDTAUFFBYTHA-UHFFFAOYSA-N
Synonym 7h-purin-6-amine hydrochloride hydrate, adenine hydrochloride hydrate, adenine hydrochloride hemihydrate, 1h-purin-6-amine monohydrochloride hydrate 2:1, ade hydrate hydrochloride, purine-6-ylamine, chloride, hydrate, c5h5n5.hcl.h2o, adenine hydrochloride, adeninehydrochloridehemihydrate, vitamin b4 hydrate hydrochloride
PubChem CID 24211900
IUPAC Name 7H-purin-6-amine hydrate hydrochloride
SMILES O.Cl.NC1=C2NC=NC2=NC=N1

Specifications

Absorbance λ max 262 nm (0.1 N HCl)
Color White
Assay Percent Range 0.992
Quantity 5 g
Formula Weight 171.6
Percent Purity 99%
Physical Form Powder
For Use With (Application) Used as a blood preservative and in viral research, Regulatory molecule
Chemical Name or Material Adenine hydrochloride

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.