Learn More
Memmert™ Additional Stainless Steel Grid, electro-polished, For Uf750Ts
Ideal for ramp operation with rapid temperature changes during incubation, breeding or storage.
Brand: Memmert™ UF750TSE20182
Additional Details : Weight : 0.01100kg
Description
- For use with compressor-cooled incubator models 450 and 750
- Grid pattern allows for airflow
- Manufactured from electro-polished stainless steel
Specifications
Models 450 and 750 Ovens | |
Grid | |
Stainless Steel |