Learn More
Cytiva ADCF-MAb Media
Product Code. 10788134
Change view
Click to view available options
Quantity:
500 mL
1000 mL
6 x 1000 mL
Unit Size:
1000mL
500mL
Pack of 6
Product Code. Quantity unitSize
10788134 500 mL 500mL
10094723 6 x 1000 mL Pack of 6
10397042 1000 mL 1000mL
3 options
Product Code. 10788134
Brand: Cytiva SH30349.01
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.
  • ADCF
  • Metabolically designed for high cell yield and monoclonal antibody production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Contains L-glutamine and Poloxamer 188
  • Formulations without L-glutamine support the GS gene expression system
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type ADCF-mAb Liquid Medium
Sterilization Method 0.2 μm Sterile-filtered
With Additives Glutamine, Poloxamer 188, HEPES, Sodium Bicarbonate
Without Additives No Protein, No Phenol Red
Quantity 500 mL
Certifications/Compliance cGMP
Content And Storage 2° to 8°C, away from light
Description with L-Glutamine