Learn More
Adavosertib, MedChemExpress
Product Code. p-200096122
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444338 10 mg 10mg
16417057 5 mg 5mg
16400667 50 mg 50mg
16415256 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16444338 Supplier Medchem Express Supplier No. HY10993
MedChemExpress Adavosertib (AZD-1775; MK-1775) is a potent Wee1 inhibitor with an IC50 of 5.2 nM.
Adavosertib (AZD-1775
Specifications
Chemical Name or Material Adavosertib
CAS 955365-80-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H32N8O2
Quantity 10 mg
Synonym AZD1775 MK-1775
Solubility Information DMSO : 125 mg/mL (249.70 mM; Need ultrasonic)
SMILES O=C1N(N(C2=NC(NC3=CC=C(C=C3)N4CCN(CC4)C)=NC=C21)C5=NC(C(O)(C)C)=CC=C5)CC=C
Molecular Weight (g/mol) 500.6
Formula Weight 500.6
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease