Learn More
Adarotene, MedChemExpress
Product Code. 16498777
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498777 5 mg 5mg
16492317 50 mg 50mg
16426188 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16498777 Supplier Medchem Express Supplier No. HY14808
MedChemExpress Adarotene is an effective apoptosis inducer, which surprisingly produces DNA damage and exhibites a potent antiproliferative activity on a large panel of human tumor cells.
Adarotene is an effective apoptosis inducer, which surprisingly produces DNA damage and exhibites a potent antiproliferative activity on a large panel of human tumor cells.
Specifications
Chemical Name or Material Adarotene
CAS 496868-77-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H26O3
Quantity 5 mg
Synonym ST1926
Solubility Information DMSO : 25 mg/mL (66.76 mM; Need ultrasonic)
SMILES O=C(O)/C=C/C1=CC=C(C2=CC=C(O)C(C3(C4)CC5CC4CC(C5)C3)=C2)C=C1
Molecular Weight (g/mol) 374.47
Formula Weight 374.47
Percent Purity 98.76%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death