Learn More
BTX Molecular Delivery Systems™ Plate Handler Adaptor
Product Code. 15417320
Product Code. 15417320 Supplier BTX Molecular Delivery Systems™ Supplier No. 450465
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Plate Handler Adaptor helps to create a safe and reliable working area for processing electroporation plates.
25 well plate, for HT100 and HT200 electroporator plate handlers
Specifications
Sterility Non-sterile
Type Adaptor for 25-well plates
For Use With (Equipment) HT-100, HT-200 plate handlers
For Use With (Application) High-Throughput electroporation