Learn More
Hettich™ Adapting frame for Carrier no. 5628
Click to view available options
For Use With (Equipment):
Carrier no. 5628
Unit Size:
Each
Description
- Adapting Frame
- Accessory for Carrier no. 5628
- Allows Carrier no. 5628 to be used with a wide range of Adapters for Hettich Rotors
Specifications
Specifications
| Type | Adapting Frame |
| For Use With (Equipment) | Carrier no. 5628 |