Learn More
Thermo Scientific™ Adapters for TX-150 Swinging Bucket Rotors
Product Code. 15195243 Shop All Thermo Scientific Products
Change view
Click to view available options
Tube Type:
100 mL Round Bottom Open-Top Tube
11 mL IVF Tube
13 mL Urine Tube
25 mL Universal Flat/Skirted Tube
3/5 mL Blood Collection Tube
5/7 mL Blood Collection Tube
50 mL Conical or Skirted Tube
Microtube
Unit Size:
Each
Pack of 4
Product Code. Tube Type unitSize
15195243 100 mL Round Bottom Open-Top Tube Pack of 4
15145263 25 mL Universal Flat/Skirted Tube Pack of 4
15165243 3/5 mL Blood Collection Tube Each
15183263 11 mL IVF Tube Pack of 4
15135243 50 mL Conical or Skirted Tube Pack of 4
15105253 5/7 mL Blood Collection Tube Pack of 4
15135263 Microtube Pack of 4
15551510 13 mL Urine Tube Pack of 4
8 options
Product Code. 15195243
Brand: Thermo Scientific™ 75005735
Assorted tube adapters
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type 100 mL Round Bottom Open-Top Tube
Capacity 100 mL
Max. Tube Diameter 45 mm
Max. Tube Length 117 mm
For Use With (Equipment) Round Buckets