Learn More
Thermo Scientific™ Adapters for TX-1000 Swinging Bucket Rotor
Product Code. 15302127
Change view
Click to view available options
Capacity:
100 x 9/10 mL
150 to 300 mL Small Blood Bags
164 x 4.5/6 mL
20 x 50 mL
4 x 1000 mL
4 x 250 mL
4 x 500 mL
4 x 6 mL Standard or 4 x 2 mL Deep-Well
4 x 750 mL
48 x 50 mL
8 x 250 mL
96 x 15 mL
For Use With (Equipment):
1000mL Bio-Bottle
15mL Conical Tube
250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle
250mL Thermo Scientific Nalgene Bottle
4.5/6mL Blood Collection Tube
500mL Corning Bottle
500mL Nalgene Bottle
50mL Double Biocontainment Vessel for 50mL Conical Tube (can be combined with ClickSeal lids)
50mL Thermo Scientific Nalgene Oak Ridge Tube
750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter
9/10mL Blood Collection Tube
Microplate carriers
Small bag/cell culture bags 4 x 2 bags (< 350mL)
Unit Size:
4 sets
Each
Pack of 2
Pack of 4
Product Code. Capacity For Use With (Equipment) unitSize
15302127 48 x 50 mL 50mL Thermo Scientific Nalgene Oak Ridge Tube Pack of 4
15151826 100 x 9/10 mL 9/10mL Blood Collection Tube Pack of 4
11781268 164 x 4.5/6 mL 4.5/6mL Blood Collection Tube 4 sets
15109645 150 to 300 mL Small Blood Bags Small bag/cell culture bags 4 x 2 bags (< 350mL) Each
15339695 4 x 1000 mL 1000mL Bio-Bottle Pack of 4
15382117 20 x 50 mL 50mL Double Biocontainment Vessel for 50mL Conical Tube (can be combined with ClickSeal lids) Pack of 4
15149485 4 x 500 mL 500mL Nalgene Bottle Pack of 4
15159485 4 x 250 mL 250mL Conical Bottle or 200mL Thermo Scientific Nunc Bottle or 175mL Thermo Scientific Nalgene Conical Bottle Pack of 4
15169485 4 x 500 mL 500mL Corning Bottle Pack of 4
15109504 4 x 6 mL Standard or 4 x 2 mL Deep-Well Microplate carriers Pack of 4
13408999 4 x 750 mL 750 mL Polypropylene Wide-Mouth Bio-Bottle and any TX-750 Round adapter Pack of 4
15290525 8 x 250 mL 250mL Thermo Scientific Nalgene Bottle Pack of 2
13419409 96 x 15 mL 15mL Conical Tube Pack of 4
13 options
Product Code. 15302127
Brand: Thermo Scientific™ 75004252
An assortment of adapters for outstanding flexibility
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type 50 mL Nalgene Oak Ridge Tube
Capacity 48 x 50 mL
Max. Tube Diameter 29.5 mm
Max. Tube Length 120 mm
For Use With (Equipment) 50mL Thermo Scientific Nalgene Oak Ridge Tube