Learn More
Thermo Scientific™ Adapters for Thermo Scientific™ MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
Shop All Thermo Scientific Products
Click to view available options
Capacity:
0.2 mL
0.25 mL
0.5 mL
Tube Type:
Microtube
PCR
Unit Size:
1 set
Each
Description
Adapters for Thermo Scientific MicroClick 30 x 2 Rotor and MicroClick 24 x 2 Rotor
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Microtube |
| Capacity | 0.25 mL |
| For Use With (Equipment) | MicroClick 30 x 2 and MicroClick 24 x 2 rotor |
| Tube Size | 0.25 mL |