Learn More
SymLine™ Filter Adapter
missing translation for 'orderingAttributeHoverText'
Closure Size:
CPC, GL14
missing translation for 'unitSize'
Each
Beschreibung
SymLine™ offers a suitable adapter for every common thread, so that you will have no problems connecting different thread types.
- Many useful accessories such as manifolds, shut-off valves, cabinet feed-throughs or earthing cables allow you to configure your SymLine™ system to your needs
- All accessories are electrostatically dissipative and approved for use in laboratories with explosion hazards
Spezifikation
Spezifikation
| Closure Size | CPC, GL14 |
| Closure Color | Black |
| Description | Exhaust Filter to Stainless Steel Coupling |
| Color | Black |
| Liner Material | HDPE |
| For Use With (Equipment) | Justrite Container |
| Material | HDPE |
| Product Type | Adapter |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur