Learn More
WTW™ Adapter for WTW™ OxiTop/OxiTop-C™ Measuring Head
GREENER_CHOICE
Product Code. 10736452
Change view
Click to view available options
Type:
Adapter OxiTop™-AD/SK
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10736452 Adapter OxiTop™-AD/SK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10736452 Supplier WTW™ Supplier No. 209016
Estimated Shipment: 09-09-2026
Specifications
Type Adapter OxiTop™-AD/SK
For Use With (Application) For all models of PF45/ and MF/45 sample and measuring bottles
For Use With (Equipment) OxiTop™/OxiTop™-C measuring heads
Includes 1 x OxiTop™ AD adapter, 1 x OxiTop™ SK screwable adapter, 1 x gasket DI45for adapter, 1 x screwing set VS45 for adapter, 2 x GK 600 L Rubber sleeves