Learn More
Hettich™ Adapter for Rotors no. 1613, 1615 and Centrifuges EBA 200/200 S/EBA 270
Click to view available options
Capacity:
4 mL
Unit Size:
Each
Description
- Rotor Adapter
- For use with Hettich Rotors no. 1613, 1615 and Centrifuges EBA 200/200 S/EBA 270
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom |
| Capacity | 4 mL |
| Max. Tube Diameter | 10 mm |
| Max. Tube Length | 88 mm |
| For Use With (Equipment) | Rotors no. 1613, 1615 and Centrifuges EBA 200/200 S/EBA 270 |
| Vessels per Adapter | 1 |
| No. of Places | 1 |