Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Socket
Click to view available options
Standard Taper Joint No:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 25/29
Unit Size:
Pack of 10
Description
- Sockets
- Acc. to DIN 12249 form E
- DIN standard
- Conical ground sockets
- Made of DURAN™ tubing
Specifications
Specifications
| Standard Taper Joint No | NS 10/19 |
| Thickness (Metric) Wall | 1.5 mm |
| Diameter (Metric) Outer Tube | 13 mm |
| Material | Duran Glass |