Learn More
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4433
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12013914
  • 1795.00 NOK / Each
Estimated Shipment: 28-06-2024
Description
  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
30 x 2.7 to 7 mL Tubes
100 mm
30
Blood/Urine, Round Bottom
12 mm
Carriers no. 4254, 4547-B, 4579-A, 4890