missing translation for 'onlineSavingsMsg'
Learn More

Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890

Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life

Brand:  Hettich™ 4440

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11435048

  • 2015.00 NOK / Each
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications

Specifications

Rotor Adapter
225 mL
137 mm
1
Conical Falcon
61 mm
Carriers no. 4254, 4547-B, 4579-A, 4890
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers