Learn More
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4430
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10215592
  • 2015.00 NOK / Each
Estimated Shipment: 28-06-2024
Description
  • Adapter for Carriers no. 4254, 4547-B, 4579-A, 4890
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
250 mL
162 mm
1
Corning, Nalgene and Nunc Large Conical Tubes
62 mm
Carriers no. 4254, 4547-B, 4579-A, 4890