Learn More
Hettich™ Adapter for Carriers no. 4254, 4547-B, 4579-A
Description
- Adapter for Carriers no. 4254, 4547-B, 4579-A
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Conical |
| Capacity | 12 x 15 mL |
| Max. Tube Diameter | 17 mm |
| Max. Tube Length | 120 mm |
| For Use With (Equipment) | Carriers no. 4254, 4547-B, 4579-A |
| Vessels per Adapter | 12 |