Learn More
Hettich™ Adapter for Carrier no. 5625-A
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4735
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11455068
  • 1720.00 NOK / Each
Estimated Shipment: 25-06-2024
Description
  • Adapter for Carrier no. 5625-A
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
50 mL
100 mm
3
Round Bottom
34 mm
Carrier no. 5625-A
3