Learn More
Hettich™ Adapter for Carrier no. 5625-A
Product Code. 10378962
Click to view available options
Capacity:
12 x 12 to 15 mL Tubes
2 x 100 mL
250 mL
3 x 85 to 100 mL Tubes
30 x 4 mL
5 x 50 mL
50 mL
6 x 30 mL
Unit Size:
Each
Product Code. 10378962
Brand: Hettich™ 4740
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 5625-A
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Conical, Skirted, Round Bottom
Capacity 5 x 50 mL
Max. Tube Diameter 29 mm
Max. Tube Length 115 mm
For Use With (Equipment) Carrier no. 5625-A
Vessels per Adapter 5