Learn More
Hettich™ Adapter for Carrier no. 5092
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 5128
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12063914
  • 1065.00 NOK / Each
Estimated Shipment: 27-06-2024
Description
  • Adapter for Carrier no. 5092
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
12 x 4 to 6 mL Tubes
82 mm
12
Round Bottom
12 mm
Carrier no. 5092