Learn More
Hettich™ Adapter for Carrier no. 4890
Click to view available options
Capacity:
42 x 1.5 to 2 mL Tubes
Unit Size:
Each
Description
- Adapter for Carrier no. 4890
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Conical |
| Capacity | 42 x 1.5 to 2 mL Tubes |
| Max. Tube Diameter | 11 mm |
| Max. Tube Length | 38 mm |
| For Use With (Equipment) | Carrier no. 4890 |
| Vessels per Adapter | 42 |