Learn More
Hettich™ Adapter for Carrier no. 4880
Product Code. 10510893
Change view
Click to view available options
Capacity:
14 x 5 to 12 mL Tubes
19 x 1.6 to 7 mL Tubes
2 x 100 mL
250 mL
290 mL
3 x 85 to 100 mL Tubes
4 x 45 to 50 mL Tubes
48 Microliter or 24 x 1.1 to 4 mL Tubes
5 x 50 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
Product Code. Capacity unitSize
10510893 48 Microliter or 24 x 1.1 to 4 mL Tubes Each
10397412 14 x 5 to 12 mL Tubes Each
10113382 5 x 50 mL Each
10615693 3 x 85 to 100 mL Tubes Each
10215542 19 x 1.6 to 7 mL Tubes Each
10483362 7 x 25 to 30 mL Tubes Each
11445088 4 x 45 to 50 mL Tubes Each
11455088 2 x 100 mL Each
11465088 250 mL Each
10268152 290 mL Each
10 options
Product Code. 10510893
Brand: Hettich™ 4830
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 4880
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Blood/Urine, Microliter, Round Bottom
Capacity 48 Microliter or 24 x 1.1 to 4 mL Tubes
Max. Tube Diameter 11 mm
Max. Tube Length 88 mm
For Use With (Equipment) Carrier no. 4880
Vessels per Adapter 24