Learn More
Hettich™ Adapter for Carrier no. 4880
Product Code. 10397412
Change view
Click to view available options
Capacity:
14 x 5 to 12 mL Tubes
19 x 1.6 to 7 mL Tubes
2 x 100 mL
250 mL
290 mL
3 x 85 to 100 mL Tubes
4 x 45 to 50 mL Tubes
48 Microliter or 24 x 1.1 to 4 mL Tubes
5 x 50 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
10397412 14 x 5 to 12 mL Tubes Each
10113382 5 x 50 mL Each
10615693 3 x 85 to 100 mL Tubes Each
10215542 19 x 1.6 to 7 mL Tubes Each
10483362 7 x 25 to 30 mL Tubes Each
11445088 4 x 45 to 50 mL Tubes Each
11455088 2 x 100 mL Each
11465088 250 mL Each
10268152 290 mL Each
10510893 48 Microliter or 24 x 1.1 to 4 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 10397412 Supplier Hettich™ Supplier No. 4839
Only null left
Add to basket
Add to basket
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 4880
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Microliter, Round Bottom, Conical, Skirted Conical
Capacity 14 x 5 to 12 mL Tubes
Max. Tube Diameter 17 mm
Max. Tube Length 120 mm
For Use With (Equipment) Carrier no. 4880
Vessels per Adapter 14