Learn More
Hettich™ Adapter for Carrier no. 4880
Click to view available options
Capacity:
14 x 5 to 12 mL Tubes
19 x 1.6 to 7 mL Tubes
2 x 100 mL
250 mL
290 mL
3 x 85 to 100 mL Tubes
4 x 45 to 50 mL Tubes
48 Microliter or 24 x 1.1 to 4 mL Tubes
5 x 50 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
Description
- Adapter for Carrier no. 4880
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Blood/Urine, Round Bottom |
| Capacity | 19 x 1.6 to 7 mL Tubes |
| Max. Tube Diameter | 13 mm |
| Max. Tube Length | 100 mm |
| For Use With (Equipment) | Carrier no. 4880 |
| Vessels per Adapter | 19 |