Learn More
Hettich™ Adapter for Carrier no. 4780
Click to view available options
Capacity:
14 x 12 mL
2 x 100 mL
24 x 1.5 to 2 mL Tubes
250 mL
35 x 1.1 to 5 mL Tubes
4 x 50 mL
400 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
Description
- Adapter for Carrier no. 4780
- For use with Hettich Rotors
- Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
| Type | Rotor Adapter |
| Tube Type | Round Bottom, Blood/Urine |
| Capacity | 35 x 1.1 to 5 mL Tubes |
| Max. Tube Diameter | 11 mm |
| Max. Tube Length | 92 mm |
| For Use With (Equipment) | Carrier no. 4780 |
| Vessels per Adapter | 35 |