Learn More
Hettich™ Adapter for Carrier no. 4780
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 4772
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10639453
  • 1635.00 NOK / Each
Estimated Shipment: 02-07-2024
Description
  • Adapter for Carrier no. 4780
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Specifications
Rotor Adapter
400 mL
136 mm
1
High Capacity Jar
84 mm
Carrier no. 4780