Learn More
Hettich™ Adapter for Carrier no. 4780
Product Code. 10515895
Click to view available options
Capacity:
14 x 12 mL
2 x 100 mL
24 x 1.5 to 2 mL Tubes
250 mL
35 x 1.1 to 5 mL Tubes
4 x 50 mL
400 mL
7 x 25 to 30 mL Tubes
Unit Size:
Each
Product Code. 10515895
Brand: Hettich™ 4768
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
  • Adapter for Carrier no. 4780
  • For use with Hettich Rotors
  • Compatible with a wide range of tubes to fit a variety of applications
Specifications
Type Rotor Adapter
Tube Type Round Bottom
Capacity 250 mL
Max. Tube Diameter 65 mm
Max. Tube Length 115 mm
For Use With (Equipment) Carrier no. 4780
Vessels per Adapter 2
No. of Places 1